Learn More
GIF Antibody (012), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90226AF594
Product Code. 18230198
  • € 617.00 / 0.10mL
Description
GIF Monoclonal specifically detects GIF in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GIF
Monoclonal
Alexa Fluor 594
50mM Sodium Borate
Rabbit
Protein A purified
RUO
2694
Store at 4°C in the dark.
IgG
Western Blot
12
Western Blot
gastric intrinsic factor (vitamin B synthesis), IFIntrinsic factor, IFMHgastric intrinsic factor, INF, TCN3
This antibody was obtained from a rabbit immunized with purified, recombinant Human GIF (P27352-1; Met1-Tyr417).
0.1 mL