missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GIGYF2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15576872
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15576872 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15576872 Supplier Invitrogen™ Supplier No. PA545586

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: DDPGRPNFEE GGPTSVGRKH EFIRSESENW RIFREEQNGE DEDGGWRLAG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIGYF2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GIGYF2
Gene Accession No. Q6Y7W6
Gene Alias 2610016F01Rik; A830080H02Rik; AI852361; AW259676; BC006835; GIGYF2; GRB10 interacting GYF protein 2; GRB10 interacting GYF protein 2; GRB10-interacting GYF protein 2; GRB10-interacting GYF protein 2; GYF2; KIAA0642; LOW QUALITY PROTEIN: PERQ amino acid-rich with GYF domain-containing protein 2; mKIAA0642; PARK11; Parkinson disease (autosomal recessive, early onset) 11; PERQ amino acid rich, with GYF domain 3; PERQ amino acid-rich with GYF domain-containing 2-like protein; PERQ amino acid-rich with GYF domain-containing protein 2; PERQ amino acid-rich with GYF domain-containing protein 2; GRB10-interacting GYF protein 2; PERQ2; PERQ3; rCG23949-like; TNRC15; trinucleotide repeat containing 15; trinucleotide repeat-containing gene 15 protein
Gene Symbols GIGYF2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GIGYF2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26058
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.