Learn More
Gilson™ PIPETMAN™ M Pipettes
Product Code. 15560500 Shop All Gilson Products
Change view
Click to view available options
Number of Channels:
1
8
12
Model:
P10mLM
P1200M
P12X10M
P12x100M
P12x1200M
P12x200M
P12x20M
P12x300M
P300M
P5000M
P8X20M
P8x100M
P8x10M
P8x1200M
P8x200M
P8x300M
Volume (Metric):
0.5 to 10 μL
1 to 10 mL
1 to 20 μL
10 to 10 μL
10 to 100 μL
10 to 300 μL
100 to 1200 μL
20 to 200 μL
20 to 300 μL
50 to 1200 μL
500 to 5000 μL
Compatible Tips:
D10 mL
D10, DL10, DF10 or DFL10 tips
D10, DL10, DF10, DFL10 tips
D1000, DF1000, D1200, DF1200
D1200
D200
D200, D300, DF100ST, DF200ST, DF300ST tips
D200, DF200, D300, DF300
D5000
DF100
DF1200
DL10, DF30ST, D200 tips
Unit Size:
Each
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Number of Channels Model Volume (Metric) Compatible Tips unitSize
15560500 8 P8x10M 0.5 to 10 μL D10, DL10, DF10, DFL10 tips Each
15570500 12 P12X10M 0.5 to 10 μL D10, DL10, DF10 or DFL10 tips Each
15580500 8 P8X20M 1 to 20 μL DL10, DF30ST, D200 tips Each
15590500 12 P12x20M 1 to 20 μL DL10, DF30ST, D200 tips Each
15500510 8 P8x200M 20 to 200 μL D200, D300, DF100ST, DF200ST, DF300ST tips Each
15510510 12 P12x200M 20 to 200 μL D200, D300, DF100ST, DF200ST, DF300ST tips Each
15520510 8 P8x300M 10 to 300 μL D10, DL10, DF10, DFL10 tips Each
15530510 12 P12x300M 10 to 300 μL D10, DL10, DF10 or DFL10 tips Each
15550490 1 P300M 20 to 300 μL D200, DF200, D300, DF300 Each
15560490 1 P1200M 100 to 1200 μL D1000, DF1000, D1200, DF1200 Each
15570490 1 P5000M 500 to 5000 μL D5000 Each
15580490 1 P10mLM 1 to 10 mL D10 mL Each
15590490 8 P8x100M 10 to 10 μL D200 Each
15500500 12 P12x100M 10 to 100 μL DF100 Each
15510500 8 P8x1200M 50 to 1200 μL D1200 Each
15520500 12 P12x1200M 50 to 1200 μL DF1200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Product Code. 15560500 Supplier Gilson™ Supplier No. F81025
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Single or multichannel. Equipped with motorized piston that virtually eliminates pipetting variability among users and minimizes RSI (Repetitive Strain Injuries) among users
  • Motorized piston with adjustable piston speed that is compatible with varying solution densities
  • Lockable volume to prevent inaccuracies
  • Color-coded push-button corresponds to matching tip packs
  • Service alert reminders
  • Unique tip holder design allows for a secure fit of most brands of tips
  • Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
  • Pipette can be used while charging eliminating bottlenecks and downtime
  • Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
  • Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
  • Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips D10, DL10, DF10, DFL10 tips
Color Red
Model P8x10M
Number of Channels 8
Product Line Pipetman
Type Pipette
Volume (Metric) 0.5 to 10 μL
Finger Style Ergonomic
Warranty 1 Year