missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GilZ Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-41210CL1
This item is not returnable.
View return policy
Description
GilZ Polyclonal antibody specifically detects GilZ in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| GilZ | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| Delta sleep-inducing peptide immunoreactor, DSIP-immunoreactive leucine zipper protein, DSIP-immunoreactive peptide, GILZDKFZp313A1123, Glucocorticoid-induced leucine zipper protein, Protein DIP, TSC22 domain family protein 3, TSC22 domain family, member 3, TSC-22 related protein, TSC-22-like protein, TSC-22-related protein, TSC-22Rimmunoreactor | |
| Antibody was raised against a 16 amino acid peptide near the amino terminus of human TSC22D3. The immunogen is located within amino acids 40 - 90 of TSC22D3. Amino Acid Squence: ENLNTDKLNSIMRQDS | |
| 0.1 mL | |
| Cancer | |
| 1831 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction