missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GIMAP5 Polyclonal Antibody

Product Code. 17187693
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17187693 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17187693 Supplier Invitrogen™ Supplier No. PA5109766

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: WGSVEEQRQQ QAELLAVIER LGREREGSFH SNDLFLDAQL LQRTGAGACQ EDYRQYQAKV EWQVEKHKQE LRENESNWAY K.

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GIMAP5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GIMAP5
Gene Accession No. Q8BWF2, Q8K3L6, Q96F15
Gene Alias D630024P16; E230026N22Rik; Gimap5; GTPase IMAP family member 5; GTPase of the immunity-associated protein 5; GTPase, IMAP family member 5; HIMAP3; Ian4; IAN-4; Ian4l1; IAN5; IAN-5; Iddm1; IMAP3; immune associated nucleotide 4 like 1; immune-associated nucleotide-binding protein 5; immunity associated protein 3; immunity-associated nucleotide 4 protein; Immunity-associated nucleotide 4-like 1 protein; immunity-associated nucleotide 5 protein; Immunity-associated protein 3; inhibitor of radiation- and OA-induced apoptosis, Irod/Ian5; IROD; lymphopenia; lyp
Gene Symbols GIMAP5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 190-270 of human GIMAP5 (NP_060854.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246774, 317757, 55340
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.