missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GIMAP6 Polyclonal Antibody

Description
Immunogen sequence: SLEDYVRETN NQALAWLDVT LARRHCGFNN RAQGEEQEAQ LRELMEKVEA IMWENEGDYY SNKAYQYTQQ NFRLKELQER QVSQGQGSED VPGEESWLEG LSQIQK Highest antigen sequence indentity to the following orthologs: Rat - 38%, Mouse - 38%.
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, IAN subfamily genes are located in a cluster at 7q36.1. Two transcript variants, one protein-coding and the other probably non-protein-coding, have been found for this gene.
Specifications
Specifications
| Antigen | GIMAP6 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GIMAP6 |
| Gene Accession No. | Q6P9H5 |
| Gene Alias | 4833419H03Rik; Gimap6; GTPase IMAP family member 6; GTPase, IMAP family member 6; IAN2; IAN-2; IAN6; IAN-6; immune associated nucleotide 2; immune associated nucleotide 6; immune-associated nucleotide 6; immune-associated nucleotide-binding protein 6; Immunity-associated nucleotide 2 protein; Immunity-associated nucleotide 6 protein; mFLJ00102; mIAN6 |
| Gene Symbols | GIMAP6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?