missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ULBP-1 Monoclonal antibody specifically detects ULBP-1 in Human samples. It is validated for Flow Cytometry, CyTOF, Neutralization
Specifications
Specifications
| Antigen | ULBP-1 |
| Applications | Flow Cytometry, CyTOF, Neutralization |
| Classification | Monoclonal |
| Clone | MM0592-10K10 |
| Conjugate | HRP |
| Dilution | Flow Cytometry, CyTOF-ready, Block/Neutralize |
| Formulation | PBS |
| Gene Alias | ALCAN-beta, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL1, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1 |
| Host Species | Mouse |
| Immunogen | Human recombinant ULBP-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?