missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIMAP8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 433.00 - € 572.00
Specifications
| Antigen | GIMAP8 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18440801
|
Novus Biologicals
NBP1-85060-25ul |
25ul |
€ 433.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18212578
|
Novus Biologicals
NBP1-85060 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GIMAP8 Polyclonal specifically detects GIMAP8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GIMAP8 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 155038 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PDISSLKNIDSEVRKHICTGPHAFLLVTPLGFYTKNDEAVLSTIQNNFGEKFFEYMIILLTRKEDLGDQDLDTFLRNSNKALYGLIQKCKNRYSAFNYRATGEEEQRQADELLEKIESMVHQNGNK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DKFZp667I133, GTPase IMAP family member 8, GTPase, IMAP family member 8, hIAN6, human immune associated nucleotide 6, IAN6, IAN9, IAN-9, IANT, immune associated nucleotide, Immune-associated nucleotide-binding protein 9, MGC129545, Protein IanT | |
| GIMAP8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title