missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GINS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100543
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GINS3.
This gene encodes a protein subunit of the GINS heterotetrameric complex, which is essential for the initiation of DNA replication and replisome progression in eukaryotes. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| GINS3 | |
| Polyclonal | |
| Unconjugated | |
| Gins3 | |
| 2700085M18Rik; AI616142; DNA replication complex GINS protein PSF3; GINS complex subunit 3; GINS complex subunit 3 (Psf3 homolog); Gins3; PSF3; RGD1308153 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 307639, 64785, 78833 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BRX5, Q9CY94 | |
| Gins3 | |
| A synthesized peptide derived from human GINS3(Accession Q9BRX5), corresponding to amino acid residues E130-Q180. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction