missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GINS4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA555146

Product Code. 15723071

  • € 572.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MTEEVDFLGQ DSDGGSEEVV LTPAELIERL EQAWMNEKFA PELLESKPEI VECVMEQLEH MEE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

The yeast heterotetrameric GINS complex is made up of Sld5, Psf1 (GINS1), Psf2 (GINS2), and Psf3 (GINS3). The formation of the GINS complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GINS4
Polyclonal
Unconjugated
GINS4
2810037c03rik; 4933405K01Rik; DNA replication complex GINS protein SLD5; DNA replication complex GINS protein SLD5, N-terminally processed; GINS complex subunit 4; GINS complex subunit 4 (Sld5 homolog); Gins4; RGD1307397; SLD5; SLD5 homolog
Rabbit
Antigen affinity chromatography
RUO
84296
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BRT9
GINS4
Recombinant protein corresponding to Human GINS4. Recombinant protein control fragment (Product #RP-93808).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.