missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIP Antibody (4), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-21617UV
This item is not returnable.
View return policy
Description
GIP Monoclonal specifically detects GIP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GIP | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GIP | |
| Synthetic human gastric inhibitory polypeptide adsorbed onto aluminum hydroxide gel. | |
| 0.1 mL | |
| Primary | |
| Cross-reacts up to 10% with PACAP and human GLP-2 but <0.1% with glucagon, GLP-1 or VIP. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2695 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction