missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIPR Antibody (591816), Allophycocyanin, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB82101APC
This item is not returnable.
View return policy
Description
GIPR Monoclonal specifically detects GIPR in Human samples. It is validated for Flow Cytometry.
Specifications
| GIPR | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| GIPR | |
| NS0 mouse myeloma cell line transfected with human GIPR (amino acids 1-466) | |
| 100 μL | |
| GPCR, Neuroscience | |
| 2696 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| 591816 | |
| Flow Cytometry | |
| gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction