Learn More
GIPR Antibody (591827), PE/Cy5.5, R&D Systems™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals MAB82102PECY55
Product Code. 18094816
  • ONLINE EXCLUSIVE PRICE
    € 630.00
    € 595.35 / 0.10mL
    Save € 34.65
    5% Off
Description
GIPR Monoclonal specifically detects GIPR in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GIPR
Monoclonal
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
GIPR
NS0 mouse myeloma cell line transfected with human GIPR (amino acids 1-466)
0.1 mL
Primary
Human
IgG1
Flow Cytometry
591827
Flow Cytometry
gastric inhibitory polypeptide receptor, GIP-R, Glucose-dependent insulinotropic polypeptide receptor, MGC126722, PGQTL2
Mouse
Protein A or G purified
GPCR, Neuroscience
2696.0
Store at 4°C in the dark. Do not freeze.