missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIRK2 Antibody (KCNJ6/7559), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24131AF405
This item is not returnable.
View return policy
Description
GIRK2 Monoclonal antibody specifically detects GIRK2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GIRK2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| BIR1Kir3.2, G protein-activated inward rectifier potassium channel 2, GIRK-2, GIRK2KATP-2, hiGIRK2, Inward rectifier K(+) channel Kir3.2, inward rectifier potassium channel KIR3.2, KATP2human G protein coupled inward rectifier potassium channel 2 (hiGIRK2)10KCNJ7KIR3.2, MGC126596, Potassium channel, inwardly rectifying subfamily J member 6, potassium inwardly-rectifying channel, subfamily J, member 6 | |
| Recombinant fragment of human GIRK2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 3763 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| KCNJ6/7559 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction