Learn More
GIT1 Antibody (924640), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8508N
Product Code. 30061446
  • € 597.00 / 0.10mL
Description
GIT1 Monoclonal antibody specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GIT1
Monoclonal
Alexa Fluor 700
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
924640
Western Blot, ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1
E. coli-derived recombinant human GIT1, Ser485-Asp636, Accession # Q9Y2X7
0.1 mL
Cell Biology, Cytoskeleton Markers, GPCR, Signal Transduction
28964
Store at 4C in the dark.
IgG2b