Learn More
GIT1 Antibody (924640), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8508MFV450
Product Code. 18520709
  • € 629.00 / 0.10mL
Description
GIT1 Monoclonal antibody specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GIT1
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
0.1 mL
Cell Biology, Cytoskeleton Markers, GPCR, Signal Transduction
28964
Store at 4C in the dark.
Western Blot, ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
924640
Western Blot, ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1
E. coli-derived recombinant human GIT1, Ser485-Asp636, Accession # Q9Y2X7
RUO
Primary
Human, Mouse, Rat
IgG2b