Learn More
GITR Ligand Rat anti-Mouse, Brilliant Violet 650, Clone: MIH44, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 742578
Description
The MIH44 monoclonal antibody specifically binds to Glucocorticoid-induced TNF-related Ligand (GITR Ligand). The GITR Ligand protein is encoded by the Tnfsf18 /( Tumor necrosis factor ligand superfamily member 18 /) gene. The GITR Ligand is a type II transmembrane protein that is expressed on endothelial cells and antigen presenting cells, including B cells, macrophage and dendritic cells. The GITR Ligand reportedly costimulates T cell activation and reverses CD4+CD25+ Treg cell-mediated suppression by interaction with its cognate receptor, GITR that is expressed by these cells.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Specifications
| GITR Ligand | |
| MIH44 | |
| Brilliant Violet 650 | |
| Tnfsf18, Gitrl,Glucocorticoid-induced-tumor necrosis factor receptor ligand | |
| GITRL/GFP-transfected L cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.