missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR Ligand/TNFSF18 Antibody (MM0312-4Q25), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11965UV
This item is not returnable.
View return policy
Description
GITR Ligand/TNFSF18 Monoclonal specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Flow Cytometry.
Specifications
| GITR Ligand/TNFSF18 | |
| Monoclonal | |
| DyLight 350 | |
| Q9UNG2 | |
| TNFSF18 | |
| Human recombinant GITR Ligand/TNFSF18 (extracellular domain) | |
| 0.1 mL | |
| Apoptosis | |
| 8995 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| MM0312-4Q25 | |
| Flow Cytometry | |
| Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction