missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR Ligand/TNFSF18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-80104
This item is not returnable.
View return policy
Description
GITR Ligand/TNFSF18 Polyclonal specifically detects GITR Ligand/TNFSF18 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GITR Ligand/TNFSF18 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 um-filtered solution in PBS, pH 7.4 | |
| TNFSF18 | |
| Recombinant human GITR Ligand/TNFSF18. | |
| 100 μg | |
| Apoptosis | |
| 8995 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Activation-inducible TNF-related ligand, AITRLMGC138237, GITRLglucocorticoid-induced TNFR-related protein ligand, Glucocorticoid-induced TNF-related ligand, hGITRLGITR ligand, TL6AITR ligand, tumor necrosis factor (ligand) superfamily, member 18, tumor necrosis factor ligand superfamily member 18 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognizes human GITR Ligand/TNFSF18. Detects a band of ∽15-17kDa by Western blot. Does not cross-react with mouse GITR Ligand/TNFSF18. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction