missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR/TNFRSF18 Antibody (DTA-1), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-26661AF532
This item is not returnable.
View return policy
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| GITR/TNFRSF18 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TNFRSF18 | |
| Mouse CD25+ CD4+ T cell line NP_683700 | |
| Primary | |
| Mouse | |
| IgG2b λ |
| Flow Cytometry | |
| DTA-1 | |
| Flow Cytometry | |
| Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
| Rat | |
| 0.1 mL | |
| 8784 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction