missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR/TNFRSF18 Antibody (RM0066-7C13), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12338MFV610
This item is not returnable.
View return policy
Description
GITR/TNFRSF18 Monoclonal antibody specifically detects GITR/TNFRSF18 in Mouse samples. It is validated for Flow Cytometry
Specifications
| GITR/TNFRSF18 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| RM0066-7C13 | |
| Flow Cytometry | |
| Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
| Mouse T cells | |
| 0.1 mL | |
| Apoptosis | |
| 8784 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction