missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GITR/TNFRSF18 Antibody (T9P4G4*D7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50337B
This item is not returnable.
View return policy
Description
GITR/TNFRSF18 Monoclonal specifically detects GITR/TNFRSF18 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| GITR/TNFRSF18 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TNFRSF18 | |
| Peptide Sequence - TEDARSCQFP | |
| 0.1 mL | |
| Apoptosis | |
| 8784 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA, Immunohistochemistry | |
| T9P4G4*D7 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Activation-inducible TNFR family receptor, AITRTNF receptor superfamily activation-inducible protein, CD357 antigen, GITR-D, GITRtumor necrosis factor receptor superfamily member 18, Glucocorticoid-induced TNFR-related protein, tumor necrosis factor receptor superfamily, member 18 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction