missing translation for 'onlineSavingsMsg'
Learn More

Givinostat hydrochloride monohydrate, MedChemExpress

Product Code. 16432337
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16432337

Brand: Medchem Express HY14842B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Givinostat hydrochloride monohydrate (ITF-2357 hydrochloride monohydrate) is a HDAC inhibitor with an IC50 of 198 and 157 nM for HDAC1 and HDAC3, respectively.

Givinostat hydrochloride monohydrate (ITF-2357 hydrochloride monohydrate) is a HDAC inhibitor with an IC50 of 198 and 157 nM for HDAC1 and HDAC3, respectively.

Specifications

Chemical Name or Material Givinostat hydrochloride monohydrate
Physical Form Solid
CAS 732302-99-7
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H30ClN3O5
For Use With (Application) Cancer-programmed cell death
Synonym ITF-2357 hydrochloride monohydrate
Solubility Information DMSO : ≥ 100 mg/mL (210.10 mM)
SMILES O=C(OCC1=CC=C2C=C(CN(CC)CC)C=CC2=C1)NC3=CC=C(C(NO)=O)C=C3.[H]Cl.O
Molecular Weight (g/mol) 475.97
Formula Weight 475.97
Percent Purity 96.13%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.