missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GKN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 180.00 - € 476.00
Specifications
| Antigen | GKN1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17750544
![]() |
Invitrogen™
PA5119133 |
200 μL |
€ 476.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17763354
|
Invitrogen™
PA5121958 |
20 μL |
€ 180.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GKN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Gastrokine 1, also known as GKN1 and CA11, is a 185 amino acid containing secreted protein belonging to the gastrokine family with a BRICHOS domain and an asn-rich region. GKN1 (also known as an antioncogene) is reported to be down-regulated in human gastric cancer tissue as compared to normal gastric mucosa. Differential display of normal and cancerous gastric tissue suggested that this protein is predominantly expressed in stomach and the expression is down-regulated to considerable frequency in cancer tissue or gastric cancer cell lines examined. This extra cellular protein is co-secreted with mucins and is a signaling growth factor with mitogenic activity and may be involved in maintaining the integrity of the gastric mucosal epithelium.Specifications
| GKN1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Rat, Pig | |
| GKN1 | |
| 18 kDa antrum mucosa protein; 2200002K21Rik; Amp18; AMP-18; Amp18; gastrokine; BRICD1; BRICHOS domain containing 1; Ca11; FOV; foveolin; gastrokine 1; gastrokine-1; Gkn1; Protein CA11; Protein CA11 homolog; UNQ489/PRO1005 | |
| GKN1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8HYA9, Q9NS71 | |
| 100502561, 297418, 56287 | |
| Recombinant protein Gastrokine 1. The antigen corresponds to amino acid range Ala22-Asn199 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title