missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GL7 (T and B Cell Activation Marker) Antibody (GL-7), Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-00361-50UG
This item is not returnable.
View return policy
Description
GL7 (T and B Cell Activation Marker) Monoclonal specifically detects GL7 (T and B Cell Activation Marker) in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| GL7 (T and B Cell Activation Marker) | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry 1:10-1:1000 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgM |
| Flow Cytometry | |
| GL-7 | |
| Biotin | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
| The immunogen for this antibody is T- and B-cell activation antigen (GL7, Ly-77). | |
| 50 μg | |
| Primary | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction