Learn More
Gerresheimer™ Threaded Brown Wide Mouth Bottles without Closure
28.41 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Use promo code "21637" to get your promotional price.
Click to view available options
Capacity (Metric):
30 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Material:
Glass
Description
- Round
- Wide Mouth
- Brown Glass
- Without closure
Specifications
Product Type | Threaded Bottle |
Capacity (Metric) | 30 mL |
Color | Brown |
Material | Glass |
No. per Pack | 126 Pack |
Mouth | Wide |
Shape | Round |