Learn More
Hirschmann™ Glass Coverslips for Haemacytometer
Click to view available options
Dimensions (L x W):
20 x 26 mm
22 x 22 mm
24 x 24 mm
Product Type:
Hemacytometer Coverslip
Unit Size:
Pack of 10
Description
- Surface ground and polished surface
- LOT number on the packaging
- Labelled with: usable until, CE pursuant to IVD 98/79 EC According to the German Calibration Regulation (Eichordnung), only officially calibrated haemocytometer glass covers may be placed on the market and used in the FRG
Specifications
Specifications
| Product Type | Hemacytometer Coverslip |
| Material | Glass |
| Thickness (Metric) | 0.4 mm |
| Dimensions (L x W) | 22 x 22 mm |
| For Use With (Equipment) | Haemocytometer |