Learn More
Brady™ Glass-Filled Nylon Miniature Circuit Breaker Lockouts
A safe and effective method for locking out European Circuit Breakers.
Brand:  Brady™ 090850
Additional Details : Weight : 0.01810kg
 View more versions of this product
Product Code. 11771945
  • € 22.70 / Each
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
  • Requires no tools to install
  • Lockout easily installs with the aid of a push button
  • Tie-bar style lockout has a thumb wheel for quick application
Specifications
Specifications
Miniature Circuit Breaker Lockouts
Glass-filled Nylon
Miniature Circuit Breaker Lockouts - Pin-Out Wide
Glass-filled Nylon
Red/Black
UL 94 HB
7 mm
1 Ea.