Learn More
WTW™ Glass Flow Through Vessel
Product Code. 10324621
Click to view available options
Type:
Flow-through Vessel
Unit Size:
Each
Product Code. 10324621
Brand: WTW™ 302730
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Flow vessels for WTW conductivity measuring cells are perfectly designed to perform measurements without the interference of carbon dioxide through the air or to examine the conductivity of solutions in closed circuits.The overpressure in the system must not exceed 0.5 bar.
TRUSTED_SUSTAINABILITY
Specifications
Type Flow-through Vessel