Learn More
Hirschmann™ Class AS Graduated Pipettes, Blue Graduation
Product Code. 11700834
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
25 mL
50 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. 11700834
Brand: Hirschmann™ 1101160
Only null left
Add to basket
Add to basket
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes, Blue Graduation come with a cotton plug end as standard.
  • Acc. to DIN EN ISO 835
  • Main point ring graduation in blue
  • Graduated to tip, zero on top, type 3
  • Conformity certified, with dated batch identification
Specifications
Calibrated Yes
Certifications/Compliance DIN EN ISO 835
Class AS
Material Polypropylene
Graduated Yes
Quantity 12 Pack
Tolerance 0.03 mL
Color Blue
Volume (Metric) 10 mL
Type Bulb Pipet