Learn More
Hirschmann™ Class AS Graduated Pipettes, Blue Graduation
Description
- Acc. to DIN EN ISO 835
- Main point ring graduation in blue
- Graduated to tip, zero on top, type 3
- Conformity certified, with dated batch identification
Specifications
Specifications
| Calibrated | Yes |
| Certifications/Compliance | DIN EN ISO 835 |
| Class | AS |
| Material | Silicone |
| Color | Blue |
| Volume (Metric) | 50 mL |
| Type | Spare Adapter |
| For Use With (Equipment) | Bibbette Pipet Filler |