Learn More
Trajan Scientific and Medical™ Glass Lined Stainless-Steel GC Tubing
Product Code. p-7114377
Click to view available options
Overall Length (Metric):
30 cm
60 cm
90 cm
180 cm
Diameter (Metric) Inner:
0.3 mm
0.4 mm
0.5 mm
0.70 mm
0.75 mm
0.8 mm
1 mm
2 mm
4 mm
9.5 mm
Unit Size:
Each
Product Code. 15258993
Brand: Trajan Scientific and Medical™ 082725
Only null left
Add to basket
Add to basket
Exhibits resistance to the harshest of conditions and to strong acids and bases
  • Ideal for HPLC columns, particularly for biological matrices
  • Temperature stable up to 500°C
Specifications
For Use With HPLC Columns, Biological Matrices
Type Tubing
Diameter (Metric) Inner 0.70 mm
Overall Length (Metric) 180 cm
Material Borosilicate Glass (Lining)
Max. Operating Temperature (Metric) Stable up to 500°C
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.