missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Glass Microfiber Filters, Grade MGB

Binder-free glass microfiber filters for analytical and gravimetric analyses and prefiltering.

Brand:  Sartorius Stedim Biotech™ FT-2-1102-210297

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15912730

  • € 995.00 / Pack of 100
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

  • Binder-free
  • Suitable for the following:
    • Liquid filtration
    • Prefiltration
    • Clarification of buffer and reagent solutions
    • Analytical sample preparation
    • Particle analysis
  • Non-sterile, bulk packed
Specifications

Specifications

140 g/m2
Sheet
1 μm
210 mL/min.
Hydrophilic
1 μm
White
Binder-free Glass Microfiber
Non-sterile
MGB
100/Pk.
210 x 297 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers