Learn More
Poulten & Graf GmbH Volac™ Glass Pasteur Pipets
Product Code. 10351552
Click to view available options
Length (Metric):
150 mm
230 mm
Sterility:
Non-sterile
Sterile
Includes:
Plugged
Unplugged
Unit Size:
Pack of 1000
Pack of 250
Product Code. 10351552
Brand: Poulten & Graf GmbH D810/VA
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these pipets for a variety of applications. Poulten Graf™ Volac™ Glass Pasteur Pipets feature a slightly heavier wall thickness to provide greater strength and reduce the risk of breakage in use or when fitting PVC teats.
  • Available with or without plug
  • Glass pipets come in 150, 230 and 270mm lengths
  • Packing materials are carefully chosen to ensure ease of use for the technician with maximum protection from breakage during transit and minimum lint or dust contamination
  • All standard pipettes can be supplied with 'sealed points' and break mark
Spécification
Certifications/Compliance BS5732,ISO7712
Material Soda Lime Glass
Sterility Sterile
Drops/mL 30 to 35 mL
Includes Plugged
Quantity 250 Pack
Length (Metric) 150 mm
Type Pasteur Pipet
Disposable Single-use