Learn More
Witeg™ Glass Petri Dish
Product Code. 10796595
Change view
Click to view available options
Diameter (Metric):
80 mm
90 mm
120 mm
150 mm
180 mm
200 mm
Height (Metric):
15 mm
20 mm
25 mm
30 mm
50 mm
Unit Size:
Each
Pack of 10
Pack of 18
Pack of 9
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Height (Metric) unitSize
10796595 120 mm 20 mm Each
10194434 200 mm 50 mm Pack of 10
10092944 150 mm 25 mm Pack of 18
10276154 180 mm 30 mm Pack of 9
10296534 90 mm 15 mm Each
10431555 80 mm 15 mm Each
10387844 200 mm 30 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10796595 Supplier Witeg™ Supplier No. 5 830 006
Only null left
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Culture your cells and see them clearly in a Witeg™ Glass Petri Dish, with accompanying lid.
  • Free from bubbles and other imperfections
  • With lid
  • Glass
Specifications
Closure Included Yes
Closure Material Borosilicate Glass
Material Glass
Shape Round
Type Petri Dish
Height (Metric) 20 mm
Diameter (Metric) 120 mm