Learn More
Hach Lange™ Glass Radiometer Analytical PHC2005-8 Combination Red-Rod pH Electrode
Product Code. 10502963
Change view
Click to view available options
Connector Types:
BNC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
10502963 BNC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10502963 Supplier Hach Lange™ Supplier No. E16M500
Only null left
Specifications
Connector Types BNC
Junction Porous Pin
Type pH Electrode
Filling Solution Type Refillable: Sat. KCl
For Use With (Equipment) Hach sension benchtop pH meters
Includes Radiometer Analytical PHC2005-8 combination pH electrode with Certificate of Conformity and small bottle of internal electrolyte solution.