Learn More
Duran™ Glass Rolled Flange Bottles with Clamp Closure
Brand:  Duran™ 214655404
Additional Details : Weight : 7.30000kg
 View more versions of this product
Product Code. 15251759
  • € 440.00 / Pack of 10
Estimated Shipment
04-06-2020
Add to basket
Specifications
Specifications
1000 mL
Porcelain Clamp Closure, Replacement Rubber Seal
10Pack
Up to 121°C
Sampling and cultivation.
Glass
Rolled Flange Bottle
95mm