Learn More
Gravis Glass Sterilizing Jars
Click to view available options
Capacity (Metric):
500 mL
750 mL
1000 mL
2000 mL
Unit Size:
Pack of 22
Pack of 36
Pack of 6
Description
- Manufactured from soda lime glass
- Available in a range of capacities
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | Soda Lime Glass |
| Quantity | 6Pack |
| Weight (Metric) | 0.8 kg |
| Shape | Round |
| Product Type | Sterilizing Jar |
| Closure Type | Cap |