Learn More
Karl Hecht™ Glass Stirring Rods
Product Code. 10378731
Click to view available options
Length (Metric):
200 mm
Diameter (Metric):
3 mm
Unit Size:
Each
Product Code. 10378731
Brand: Karl Hecht™ 42850024
Only null left
Add to basket
Add to basket
An excellent option for the mixing and stirring of chemicals and other liquids in the laboratory.
  • With fused ends
  • Available in a range of lengths and diameters
  • General laboratory use
Specifications
Description Fused ends
For Use With (Application) Stirring, mixing chemicals and liquids
Diameter (Metric) 3 mm
Length (Metric) 200 mm
Material Glass
Type Stirring Rod