Learn More
Karl Hecht™ Glass Stirring Rods
Click to view available options
Length (Metric):
200 mm
Diameter (Metric):
3 mm
Unit Size:
Each
Description
- With fused ends
- Available in a range of lengths and diameters
- General laboratory use
Specifications
Specifications
| Description | Fused ends |
| For Use With (Application) | Stirring, mixing chemicals and liquids |
| Diameter (Metric) | 3 mm |
| Length (Metric) | 200 mm |
| Material | Glass |
| Type | Stirring Rod |