Learn More
DWK Life Sciences Glass Stopper, Clear, SBW Glass, Short Standard Taper Ground Joint, Octagonal flat head
Clear SBW glass ground stopper, short-length
Brand: DWK Life Sciences 246250802
Additional Details : Weight : 0.45000kg
Description
Solid or semi-hollow, short-length, clear SBW glass stoppers can be used to close glassware items such as flasks or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head.
- All glass components and therefore suitable for storage of aggressive materials
- Tight closure due to standard ground joint
- Octagonal shaped head
Specifications
Solid | |
10/Pk. | |
ST 24/20 |
Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders | |
SBW Glass | |
No |