Learn More
Hirschmann™ Class A Volumetric Flasks with Rim
Product Code. 11788061
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Unit Size:
Each
Pack of 2
Product Code. 11788061
Brand: Hirschmann™ 2800186
Only null left
Add to basket
Add to basket
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with Rim are manufactured from durable DURAN™ glass tubing.
  • With blue graduation
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/Compliance DIN EN ISO 1042
Capacity (Metric) 250 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 220 mm
Limit of Error ±0.15 mL
Tolerance ±0.15 mL
Graduated Yes
Class A