Learn More
Hofmann Klaus™ Glass Weighing Boats With Stem
Click to view available options
Capacity (Metric):
3 mL
6 mL
10 mL
Unit Size:
Each
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 10 mL |
| Material | Glass |
| Shape | Boat |
| Type | Weighing Boat |
| For Use With (Application) | Ideal for lossless pouring of the sample |
| Size | 3 |
| Length (Metric) | 80 mm |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion