Learn More
Hofmann Klaus™ Glass Weighing Boats With Stem
Click to view available options
Capacity (Metric):
3 mL
6 mL
10 mL
Unit Size:
Each
Specifications
Specifications
| Bottom Shape | Flat |
| Capacity (Metric) | 10 mL |
| Material | Glass |
| Shape | Boat |
| Type | Weighing Boat |
| For Use With (Application) | Ideal for lossless pouring of the sample |
| Size | 3 |
| Length (Metric) | 80 mm |