missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564417
This item is not returnable.
View return policy
Description
Immunogen sequence: GRYWPARGPQ LTLFVPQHIL MTSAPNTITV LELEWAPCSS DDPELCAVTF VDRPVIGSSV TYDHPSKPVE KRLMPPPPQK NKDSWLDHV Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 46%.
Beta Galactosidase encodes a member of the glycosyl hydrolase 35 family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme. This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates. Mutations in this gene have been associated with GM1-gangliosidosis and Morquio B syndrome. Diseases associated with GLB1 include Gm1-Gangliosidosis, Type I and Gm1-Gangliosidosis, Type Ii.
Specifications
| GLB1 | |
| Polyclonal | |
| Unconjugated | |
| GLB1 | |
| acid beta-galactosidase; AW125515; betagal; Beta-Gal; Beta-galactosidase; bgal; Bge; Bgl; Bgl-e; Bgl-s; Bgl-t; Bgs; Bgt; C130097A14Rik; EBP; Elastin receptor 1; elastin receptor 1, 67kDa; ELNR1; galactosidase beta 1; galactosidase, beta 1; GLB1; Glb-1; lactase; lysosmal hydrolase; lysosomal beta-galactosidase; lysosomal hydrolase; MPS4B; βgal; β-gal; β-galactosidase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2720 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.29 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P16278 | |
| GLB1 | |
| Recombinant protein corresponding to Human beta Galactosidase | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction