missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLB1L2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15780802 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15780802 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15780802 Supplier Invitrogen™ Supplier No. PA563023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RGRVNYGENI DDQRKGLIGN LYLNDSPLKN FRIYSLDMKK SFFQRFGLDK WSSL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 80%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GLB1L2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLB1L2
Gene Accession No. Q8IW92
Gene Alias Beta-galactosidase-1-like protein 2; galactosidase beta 1 like 2; galactosidase beta 1-like 2; galactosidase, beta 1-like 2; GLB1L2; MST114; MSTP014; MSTP114; UNQ210/PRO236
Gene Symbols GLB1L2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLB1L2. Recombinant protein control fragment (Product #RP-101367).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89944
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.