missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLDC Polyclonal Antibody
Description
GLDC Polyclonal Antibody for Western Blot
Degradation of glycine is brought about by the glycine cleavage system, which is composed of four mitochondrial protein components: P protein (a pyridoxal phosphate-dependent glycine decarboxylase), H protein (a lipoic acid-containing protein), T protein (a tetrahydrofolate-requiring enzyme), and L protein (a lipoamide dehydrogenase). The protein is the P protein, which binds to glycine and enables the methylamine group from glycine to be transferred to the T protein. Defects in this gene are a cause of nonketotic hyperglycinemia (NKH).
Specifications
Specifications
| Antigen | GLDC |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GLDC |
| Gene Accession No. | P23378, Q91W43 |
| Gene Alias | D030049L12Rik; D19Wsu57e; GCE; GCSP; GLDC; Glycine cleavage system P protein; glycine cleavage system protein P; glycine decarboxylase; glycine decarboxylase P-protein; glycine decarboxylase, glycine cleavage system protein P); glycine dehydrogenase (aminomethyl-transferring); glycine dehydrogenase (decarboxylating); glycine dehydrogenase (decarboxylating), mitochondrial; glycine dehydrogenase (decarboxylating; glycine decarboxylase, glycine cleavage system protein P); glycine dehydrogenase [decarboxylating], mitochondrial; HYGN1; RP11-472F14.1 |
| Gene Symbols | GLDC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?