missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI-2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-874AF405
This item is not returnable.
View return policy
Description
GLI-2 Polyclonal antibody specifically detects GLI-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow
Specifications
| GLI-2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry | |
| Alexa Fluor 405 | |
| GLI family zinc finger 2, GLI-Kruppel family member GLI2, glioma-associated oncogene family zinc finger 2, HPE9tax helper protein 1, oncogene GLI2, Tax helper protein, tax helper protein 2, tax-responsive element-2 holding protein, tax-responsive element-25-bp sequence binding protein, THP, THP1, THP2, zinc finger protein GLI2 | |
| This affinity purified GLI-2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near amino acids 30-65 of human GLI-2 (isoform a). (Uniprot: P10070) | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Cell Cycle and Replication, Neuroscience, Stem Cell Signaling Pathway | |
| 2736 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction