missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI-3 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79019V
This item is not returnable.
View return policy
Description
GLI-3 Polyclonal antibody specifically detects GLI-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GLI-3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ACLSPAPAPHS, GCPS, GLI family zinc finger 3, GLI3 form of 190 kDa, GLI3 full length protein, GLI3-190, GLI3FL, GLI-Kruppel family member GLI3, glioma-associated oncogene family zinc finger 3, Greig cephalopolysyndactyly syndrome, oncogene GLI3, PAP-A, PAPA1, PAPBDNA-binding protein, PPDIV, zinc finger protein GLI3 | |
| The immunogen recognized by this antibody maps to a region between residue 875 and 925 of human GLI Family Zinc Finger 3 using the numbering given in entry NP_000159.3 (GeneID 2737). | |
| 0.1 mL | |
| Cancer, Stem Cell Signaling Pathway | |
| 2737 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction