missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI1 Monoclonal Antibody (OTI12D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803970S
This item is not returnable.
View return policy
Description
GLI1 Monoclonal antibody specifically detects GLI1 in Human samples. It is validated for Western BlotSpecifications
| GLI1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08151 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2735 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI12D5 | |
| Unconjugated | |
| GLI1 | |
| AV235269; GLI; GLI family zinc finger 1; Gli1; GLI-Kruppel family member GLI1; Glioma-associated oncogene; glioma-associated oncogene 1; glioma-associated oncogene homolog; glioma-associated oncogene homolog 1 (zinc finger protein); oncogene GLI; Zfp5; Zfp-5; zinc finger protein 5; zinc finger protein GLI1 | |
| Human recombinant protein fragment corresponding to amino acids 755-1106 of human GLI1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction